Ethanolamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2-Aminoethanol, Monoethanolamine
M-Clarity™ quality level = MQ200
02400-1L
02400-250ML
SIAL02400-1LEA
78.5
EUR
SIAL02400-1L
SIAL02400-250ML
Ethanolamine, Sigma-Aldrich®
Ethanolamine
Formula:
NH₂CH₂CH₂OH Boiling Pt: 171 °C (1013 hPa) Melting Pt: 10.5 °C Density: 1.0164 g/cm³ (20 °C) Flash Pt: 93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008183 CAS Number: 141-43-5 EINECS: 205-483-3 UN: 2491 ADR: 8,III Merck Index: 13,03762 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...