Ethanolamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Aminoethanol, Monoethanolamine
M-Clarity™ quality level = MQ200
411000-100ML 411000-1L
SIAL411000-100MLEA 57.8 EUR
SIAL411000-100ML SIAL411000-1L
Ethanolamine, Sigma-Aldrich®
Ethanolamine
Formula: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Boiling Pt: 171
Melting Pt: 10.5
Density: 1.0164 g/cm³ (20 °C)
Flash Pt: 93
Storage Temperature: Ambient
MDL Number: MFCD00008183
CAS Number: 141-43-5
EINECS: 205-483-3
UN: 2491
ADR: 8 III
Merck Index: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM