Ethanolamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
2-Aminoethanol, Monoethanolamine
M-Clarity™ quality level = MQ300
E9508-100ML
E9508-1L
E9508-2.5L
E9508-500ML
SIALE9508-100MLEA
35.7
EUR
SIALE9508-100ML
SIALE9508-1L
SIALE9508-2.5L
SIALE9508-500ML
Ethanolamine, Sigma-Aldrich®
Ethanolamine
Formula:
NH₂CH₂CH₂OH MW: 61.08 g/mol Boiling Pt: 171 Melting Pt: 10.5 Density: 1.0164 g/cm³ (20 °C) Flash Pt: 93 Storage Temperature: Ambient |
MDL Number:
MFCD00008183 CAS Number: 141-43-5 EINECS: 205-483-3 UN: 2491 ADR: 8 III Merck Index: 13,03762 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...