Dibutylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Di-n-butylamine, N-butyl-1-propanamine
M-Clarity™ quality level = MQ200
471232-100ML
471232-18L
471232-1L
471232-500ML
SIAL471232-100MLEA
36.6
EUR
SIAL471232-100ML
SIAL471232-18L
SIAL471232-1L
SIAL471232-500ML
Dibutylamine, Sigma-Aldrich®
Dibutylamine
Formula:
C₈H₁₉N MW: 129.25 g/mol Boiling Pt: 159 Melting Pt: –62 °C Density: 0.759 g/cm³ (20 °C) Flash Pt: 43 Storage Temperature: Ambient |
MDL Number:
MFCD00009429 CAS Number: 111-92-2 EINECS: 203-921-8 UN: 2248 ADR: 8 II Merck Index: 13,03058 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...