Spermidine, Sigma-Aldrich®
Supplier: Merck
85558-1G
85558-5G
SIAL85558-1GEA
59.6
EUR
SIAL85558-1G
SIAL85558-5G
Spermidine, Sigma-Aldrich®
Spermidine
Formula:
C₇H₁₉N₃ Boiling Pt: 246 °C (1013 hPa) Melting Pt: 20…25 °C Density: 0.925 g/cm³ (20 °C) |
MDL Number:
MFCD00008229 CAS Number: 124-20-9 EINECS: 204-689-0 Merck Index: 12,08893 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...