2-Methylaminoethanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-Methyl-2-ethanolamin, N-Methyl-2-aminoethanol, 2-(Methylamino)ethanol
M-Clarity™ quality level = MQ200
471445-25ML 471445-250ML 471445-2L
SIAL471445-25MLEA 19.6 EUR
SIAL471445-25ML SIAL471445-250ML SIAL471445-2L
2-Methylaminoethanol, Sigma-Aldrich®
2-Methylaminoethanol
Formula: C₃H₉NO
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: –3 °C
Density: 0.937 g/cm³ (20 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD00002839
CAS Number: 109-83-1
EINECS: 203-710-0
Merck Index: 12,06096