2-(Cyclohexylamino)ethanesulphonic acid (CHES)

Supplier: Roth Carl

Synonyms: N-Cyclohexyltaurine, CHES

9166.1 9166.2 9166.3
ROTH9166.1EA 0 EUR
ROTH9166.1 ROTH9166.2 ROTH9166.3
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
2-(Cyclohexylamino)ethanesulphonic acid (CHES)
Formula: C₈H₁₇NO₃S
MW: 207.29 g/mol
Melting Pt: 320
Storage Temperature: Ambient
MDL Number: MFCD00003835
CAS Number: 103-47-9
EINECS: 203-115-6
Merck Index: 13,02063

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR