Dipropyl ether, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
111333-100G
111333-25G
SIAL111333-100GEA
226
EUR
SIAL111333-100G
SIAL111333-25G
Dipropyl ether, Sigma-Aldrich®
Dipropyl ether
Formula:
C₆H₁₄O Boiling Pt: 89 °C (1013 hPa) Melting Pt: –122 °C Density: 0.736 g/cm³ (20 °C) Flash Pt: –28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009376 CAS Number: 111-43-3 EINECS: 203-869-6 UN: 2384 ADR: 3,II Merck Index: 12,08042 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...