Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99% stabilised, ReagentPlus®, Reagent Grade
Supplier: Merck
Danger
Synonyms: Ethylene glycol monomethyl ether
Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 50 ppm
185469-1L 185469-500ML
SIAL185469-1LEA 71.5 EUR
SIAL185469-1L SIAL185469-500ML
Ethylene glycol monomethyl ether (2-methoxyethanol) ≥99% stabilised, ReagentPlus®, Reagent Grade
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
Boiling Pt: 124.6 °C (1013 hPa)
Melting Pt: -85 °C
Density: 0.964 g/cm³ (20 °C)
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III
Merck Index: 12,06118
New Product