Butyric anhydride, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

B103551-1L B103551-500ML
SIALB103551-1LEA 68.9 EUR
SIALB103551-1L SIALB103551-500ML
Butyric anhydride, Sigma-Aldrich®
Butyric anhydride
Formula: C₈H₁₄O₃
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –73 °C
Density: 0.963 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00009389
CAS Number: 106-31-0
EINECS: 203-383-4
UN: 2739
ADR: 8,III
Merck Index: 13,01594

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR