Butyric anhydride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ300
W527211-1KG
W527211-25KG
W527211-SAMPLE
SIALW527211-1KGEA
114
EUR
SIALW527211-1KG
SIALW527211-25KG
SIALW527211-SAMPLE
Butyric anhydride, Sigma-Aldrich®
Butyric anhydride
Formula:
C₈H₁₄O₃ Boiling Pt: 198 °C (1013 hPa) Melting Pt: –73 °C Density: 0.963 g/cm³ (20 °C) Flash Pt: 83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009389 CAS Number: 106-31-0 EINECS: 203-383-4 UN: 2739 ADR: 8,III Merck Index: 13,01594 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...