Acetic anhydride
Supplier: Roth Carl
|
Danger
|
4483.1
4483.2
ROTH4483.1EA
43.2
EUR
ROTH4483.1
ROTH4483.2
Acetic anhydride
Acetic anhydride
Formula:
(CH₃CO)₂O Boiling Pt: 139.9 °C (1013 hPa) Melting Pt: –73 °C Density: 1.08 g/cm³ (20 °C) Flash Pt: 49 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008705 CAS Number: 108-24-7 EINECS: 203-564-8 UN: 1715 ADR: 8,II REACH: 01-2119486470-36 Merck Index: 13,00057 |