Acetic anhydride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Acetyl oxide, Acetyl acetate
M-Clarity™ quality level = MQ100
691275-200KG 691275-25KG 691275-5KG
SIAL691275-200KGEA 4370 EUR
SIAL691275-200KG SIAL691275-25KG SIAL691275-5KG
Acetic anhydride, Sigma-Aldrich®
Acetic anhydride
Formula: (CH₃CO)₂O
MW: 102.09 g/mol
Boiling Pt: 139.9
Melting Pt: –73 °C
Density: 1.08 g/cm³ (20 °C)
Flash Pt: 49
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8 II
REACH: 01-2119486470-36
Merck Index: 13,00057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f107a66d926eb36bee4d132880765bc7fc2db7c8&vid=1c6b77698bd69c2f562bcda9bf02b8dc694a0d16&prid=web&clid=SAPDAM