Caproic anhydride ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hexanoic anhydride

B23756.0B B23756.18 B23756.30 B23756.C4
B23756.0BEA 254 EUR
B23756.0B B23756.18 B23756.30 B23756.C4
Caproic anhydride ≥98%
Caproic anhydride

Purity: 98% CAS No.: 2051-49-2 Molecular Formula: C12H22O3 Formula Weight: 214.31 Flash Point: 128°(262°F) Refractive Index: 1.4290 Odour: Unpleasant Melting Point: -40° Boiling Point: 246-248° Beilstein Reference: 1776561 UN No.: 3265 MDL No.: MFCD00009509

Formula: C₁₂H₂₂O₃
Boiling Pt: 246…248 °C
Melting Pt: –40 °C
Density: 0.928
Flash Pt: 128 °C (262 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009509
CAS Number: 2051-49-2
EINECS: 218-121-4
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR