1-Dodecanol, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Undecyl carbinol, 1-Dodecyl alcohol, 1-Hydroxy dodecane, Lauryl alcohol, Dodecyl Alcohol

M-Clarity™ quality level = MQ300

W261718-1KG-K W261718-20KG-K W261718-4KG-K W261718-8KG-K
SIALW261718-1KG-KEA 74.7 EUR
SIALW261718-1KG-K SIALW261718-20KG-K SIALW261718-4KG-K SIALW261718-8KG-K
1-Dodecanol, Sigma-Aldrich®
1-Dodecanol
Formula: H₃C(CH₂)₁₀CH₂OH
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 22…24 °C
Density: 0.81976 g/cm³ (20 °C)
Flash Pt: 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004753
CAS Number: 112-53-8
EINECS: 203-982-0
UN: 3077
ADR: 9,III
Merck Index: 13,03439

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR