1-Hexadecanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Cetyl alcohol, Palmityl alcohol
M-Clarity™ quality level = MQ300
W255408-10KG W255408-1KG W255408-SAMPLE
SIALW255408-10KGEA 427 EUR
SIALW255408-10KG SIALW255408-1KG SIALW255408-SAMPLE
1-Hexadecanol, Sigma-Aldrich®
1-Hexadecanol
Formula: H₃C(CH₂)₁₄CH₂OH
Boiling Pt: 305…330 °C (1013 hPa)
Melting Pt: 46…52 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00004760
CAS Number: 36653-82-4
EINECS: 253-149-0
Merck Index: 13,02037