1-Octanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: n-Octanol
M-Clarity™ quality level = MQ100
297887-100ML 297887-1L
SIAL297887-100MLEA 116 EUR
SIAL297887-100ML SIAL297887-1L
1-Octanol, Sigma-Aldrich®
1-Octanol
Formula: H₃C(CH₂)₆CH₂OH
MW: 130.23 g/mol
Boiling Pt: 195
Melting Pt: –16 °C
Density: 0.8246 g/cm³ (20 °C)
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00002988
CAS Number: 111-87-5
EINECS: 203-917-6
Merck Index: 13,06784
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7db4a6be3aedee4af43781ec6deb31631bc376c1&vid=80cb13f618a2d3181daf8310324993f1f7c69fb2&prid=web&clid=SAPDAM