1-Pentanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol
M-Clarity™ quality level = MQ300
W205605-1KG W205605-20KG W205605-4KG W205605-8KG W205605-SAMPLE
SIALW205605-1KGEA 67.4 EUR
SIALW205605-1KG SIALW205605-20KG SIALW205605-4KG SIALW205605-8KG SIALW205605-SAMPLE
1-Pentanol, Sigma-Aldrich®
1-Pentanol
Formula: H₃C(CH₂)₃CH₂OH
MW: 88.15 g/mol
Boiling Pt: 137.9
Melting Pt: –79 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00002977
CAS Number: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3 III
Merck Index: 13,07195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92dac1192eb8e068fc1804fdcf488129a2385918&vid=d4c7f2c5672a97c6dfaddc50807d768068096252&prid=web&clid=SAPDAM