Sodium methoxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sodium methylate, Sodium methanolate
M-Clarity™ quality level = MQ200
164992-100G 164992-500G 164992-5G
SIAL164992-100GEA 51.9 EUR
SIAL164992-100G SIAL164992-500G SIAL164992-5G
Sodium methoxide, Sigma-Aldrich®
Sodium methoxide
Formula: CH₃NaO
MW: 54.02 g/mol
Melting Pt: 127
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00012179
CAS Number: 124-41-4
EINECS: 204-699-5
UN: 1431
ADR: 4.2 II