1,8-Octanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,8-Dihydroxy octane
MQ100
O3303-100G O3303-25G
SIALO3303-100GEA 115 EUR
SIALO3303-100G SIALO3303-25G
1,8-Octanediol, Sigma-Aldrich®
1,8-Octanediol
Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 279
Melting Pt: 58…61 °C
Density: 0.941 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002989
CAS Number: 629-41-4
EINECS: 211-090-8