Dithiothreitol (DTT, Cleland's reagent), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol, (R*,R*)-1,4-Dimercapto-2,3-butandiol, DTT, DL-1,4-Dithiothreitol
M-Clarity™ quality level = MQ200
D0632-1G D0632-100G D0632-10G D0632-25G D0632-5G
SIALD0632-1GEA 51 EUR
SIALD0632-1G SIALD0632-100G SIALD0632-10G SIALD0632-25G SIALD0632-5G
Dithiothreitol (DTT, Cleland's reagent), Sigma-Aldrich®
Dithiothreitol (DTT, Cleland's reagent)
Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: ∼125 °C (1013 hPa)
Melting Pt: 42…44 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004877
CAS Number: 3483-12-3
EINECS: 222-468-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5022472c3f3baef75a703791c548fc6504df810f&vid=81529732db015cb66c1f106092309ae5714325b7&prid=web&clid=SAPDAM