Diethyl sulphate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
D100706-100G D100706-1KG D100706-2KG
SIALD100706-100GEA 51.2 EUR
SIALD100706-100G SIALD100706-1KG SIALD100706-2KG
Diethyl sulphate, Sigma-Aldrich®
Diethyl sulphate
Formula: C₄H₁₀O₄S
Boiling Pt: 208 °C (1013 hPa)
Melting Pt: –25 °C
Density: 1.179 g/cm³ (20 °C)
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00009099
CAS Number: 64-67-5
EINECS: 200-589-6
UN: 1594
ADR: 6.1,II
Merck Index: 13,03156