Sodium dodecyl sulphate (SDS), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
M-Clarity™ quality level = MQ200
L5750-100G L5750-1KG L5750-500G L5750-5KG
SIALL5750-100GEA 24.7 EUR
SIALL5750-100G SIALL5750-1KG SIALL5750-500G SIALL5750-5KG
Sodium dodecyl sulphate (SDS), Sigma-Aldrich®
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM