1-Butanethiol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Mercaptobutane, n-Butanethiol, NBM, n-Butyl mercaptan
M-Clarity™ quality level = MQ200
112925-1L 112925-250ML
SIAL112925-1LEA 141 EUR
SIAL112925-1L SIAL112925-250ML
1-Butanethiol, Sigma-Aldrich®
1-Butanethiol
Formula: C₄H₁₀S
Boiling Pt: 98 °C (1013 hPa)
Melting Pt: –116 °C
Density: 0.841 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004905
CAS Number: 109-79-5
EINECS: 203-705-3
UN: 2347
ADR: 3,II
Merck Index: 12,01611