1-Dodecanethiol ≥98%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Mercaptan C12, 1-Mercaptododecane, n-Dodecanethiol, Lauryl mercaptan, n-Dodecyl mercaptan, NDM
M-Clarity™ quality level = MQ200
471364-100ML 471364-18L 471364-2L 471364-500ML
SIAL471364-100MLEA 40.9 EUR
SIAL471364-100ML SIAL471364-18L SIAL471364-2L SIAL471364-500ML
1-Dodecanethiol ≥98%, Sigma-Aldrich®
1-Dodecanethiol
Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 269…273 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0.855 g/cm³ (20 °C)
Flash Pt: 84
Storage Temperature: Ambient
MDL Number: MFCD00004885
CAS Number: 112-55-0
EINECS: 203-984-1
UN: 1760
ADR: 8 III