1-Octanethiol, Sigma-Aldrich®
Supplier: Merck
Synonyms: n-Octyl mercaptan, 1-Mercaptooctane, n-Octanethiol
M-Clarity™ quality level = MQ200
471836-250ML 471836-25ML 471836-2L
SIAL471836-250MLEA 60 EUR
SIAL471836-250ML SIAL471836-25ML SIAL471836-2L
1-Octanethiol, Sigma-Aldrich®
1-Octanethiol
Formula: C₈H₁₈S
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: –49 °C
Density: 0.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004912
CAS Number: 111-88-6
EINECS: 203-918-1
UN: 3082
ADR: 9,III