1-Propanethiol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: n-Propanethiol, Propyl mercaptan, n-Propyl mercaptan, 1-Mercaptopropane

M-Clarity™ quality level = MQ300

W352101-1KG-K W352101-2KG-K
SIALW352101-1KG-KEA 168 EUR
SIALW352101-1KG-K SIALW352101-2KG-K
1-Propanethiol, Sigma-Aldrich®
1-Propanethiol
Formula: C₃H₈S
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: –113 °C
Density: 0.82995 g/cm³ (20 °C)
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00004900
CAS Number: 107-03-9
EINECS: 203-455-5
UN: 2402
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR