2-Propanethiol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Isopropyl mercaptan, Isopropanethiol, 2-Mercaptopropane
M-Clarity™ quality level = MQ100
P50773-100ML P50773-500ML
SIALP50773-100MLEA 55.4 EUR
SIALP50773-100ML SIALP50773-500ML
2-Propanethiol, Sigma-Aldrich®
2-Propanethiol
Formula: C₃H₈S
Boiling Pt: 57 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.81 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004863
CAS Number: 75-33-2
EINECS: 200-861-4
UN: 2402
ADR: 3,II