1-Heptanesulphonic acid sodium salt
Supplier: Molekula
Synonyms: Sodium heptane-1-sulphonate, Sodium 1-heptanesulphonate
50893677-100G 50893677-25G
MOLE50893677-100GEA 222 EUR
MOLE50893677-100G MOLE50893677-25G
1-Heptanesulphonic acid sodium salt
1-Heptanesulphonic acid sodium salt
Formula: H₃C(CH₂)₅CH₂SO₃Na
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007543
CAS Number: 22767-50-6
EINECS: 245-210-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=95c09a5691140bab7446d88919f01593e8f0b9b2&vid=7c237cde5db01d0567ff19964ca3287434747480&prid=web&clid=SAPDAM