n-Tridecane, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
T57401-100G T57401-25G
SIALT57401-100GEA 109 EUR
SIALT57401-100G SIALT57401-25G
n-Tridecane, Sigma-Aldrich®
n-Tridecane
Formula: H₃C(CH₂)₁₁CH₃
Boiling Pt: 235.4 °C (1013 hPa)
Melting Pt: –5.5 °C
Density: 0.7564 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00008979
CAS Number: 629-50-5
EINECS: 211-093-4
UN: 3295
ADR: 3,III