Polyethylene glycol diacrylate (PEGDA), M.W. 700, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: PEG hydrogel, Polyethylene glycol diacrylate, Poly(ethylene glycol) diacrylate, PEG diacrylate, PEGDA, PEG bisacrylate
M-Clarity™ quality level = MQ200
455008-500ML 455008-100ML
SIAL455008-500MLEA 169 EUR
SIAL455008-500ML SIAL455008-100ML
Polyethylene glycol diacrylate (PEGDA), M.W. 700, Sigma-Aldrich®
Polyethylene glycol diacrylate (PEGDA)
Cross-linking reagent.
  • Linear and homobifunctional
  • Contains 100 ppm MEHQ and 300 ppm BHT as inhibitor
Ω- and α- end acrylates. Suitable for polymerisation reactions.
Formula: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Melting Pt: 12…17 °C
Density: 1.12 g/ml (25 °C)
Flash Pt: 230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081876
CAS Number: 26570-48-9
Order Now
Specification Test Results
Refractive Index n20/D 1.47