Propane

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

295655-300G 295655-100G
SIAL295655-300GEA 0 EUR
SIAL295655-300G SIAL295655-100G
Propane
Propane
Formula: H₃CCH₂CH₃
Boiling Pt: –42 °C (1013 hPa)
Melting Pt: –188 °C
Density: 0.515 g/cm³ (10 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009359
CAS Number: 74-98-6
EINECS: 200-827-9
UN: 1978
ADR: 2,
Merck Index: 12,07982

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR