2-Bromo-2-methylbutane, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

306878-100G 306878-5G
SIAL306878-100GEA 167 EUR
SIAL306878-100G SIAL306878-5G
2-Bromo-2-methylbutane, Sigma-Aldrich®
2-Bromo-2-methylbutane
Formula: C₅H₁₁Br
Boiling Pt: 108 °C (1013 hPa)
Density: 1.2439 g/cm³ (0 °C)
MDL Number: MFCD00000127
CAS Number: 507-36-8
EINECS: 208-071-1
UN: 1993
ADR: 3,II
Merck Index: 12,00644

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR