(±)-2-Methylvaleraldehyde, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
258563-100ML
258563-500ML
SIAL258563-100MLEA
59.3
EUR
SIAL258563-100ML
SIAL258563-500ML
(±)-2-Methylvaleraldehyde, Sigma-Aldrich®
(±)-2-Methylvaleraldehyde
Formula:
C₆H₁₂O Boiling Pt: 119 °C (1013 hPa) Melting Pt: –100 °C Density: 0.808 g/cm³ (20 °C) |
MDL Number:
MFCD00006986 CAS Number: 123-15-9 EINECS: 204-605-2 UN: 2367 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...