Isovaleraldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Methylbutyraldehyde
M-Clarity™ quality level = MQ100
146455-100ML 146455-2.5L 146455-25ML 146455-500ML
SIAL146455-100MLEA 64.7 EUR
SIAL146455-100ML SIAL146455-2.5L SIAL146455-25ML SIAL146455-500ML
Isovaleraldehyde, Sigma-Aldrich®
Isovaleraldehyde
Formula: C₅H₁₀O
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –60 °C
Density: 0.8 g/cm³ (20 °C)
Flash Pt: 0 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007014
CAS Number: 590-86-3
EINECS: 209-691-5
UN: 2058
ADR: 3,II
Merck Index: 13,05250