3,3-Dimethylbutanone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone
M-Clarity™ quality level = MQ100
P45605-100ML P45605-500ML
SIALP45605-100MLEA 115 EUR
SIALP45605-100ML SIALP45605-500ML
3,3-Dimethylbutanone, Sigma-Aldrich®
3,3-Dimethylbutanone
Formula: C₆H₁₂O
Boiling Pt: 106 °C (1013 hPa)
Melting Pt: –53 °C
Density: 0.803 g/cm³ (20 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008846
CAS Number: 75-97-8
EINECS: 200-920-4
UN: 1993
ADR: 3,II
Merck Index: 12,07594