4-Methyl-2-pentanone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
M-Clarity™ quality level = MQ300
W273104-1KG-K W273104-8KG-K
SIALW273104-1KG-KEA 77.9 EUR
SIALW273104-1KG-K SIALW273104-8KG-K
4-Methyl-2-pentanone, Sigma-Aldrich®
4-Methyl-2-pentanone
Formula: (CH₃)₂CHCH₂COCH₃
Boiling Pt: 117.4 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.801 g/cm³ (20 °C)
Flash Pt: 16 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d44947ef43cbc8103ae523ce5da380ef184ba09&vid=373b9075524004b95d32d3cf9698896714467e8c&prid=web&clid=SAPDAM