2-Ethylcaproic acid ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A12644.0D
A12644.0F
A12644.AE
A12644.AP
A12644.0DEA
239
EUR
A12644.0D
A12644.0F
A12644.AE
A12644.AP
2-Ethylcaproic acid ≥99%
2-Ethylcaproic acid
Purity: 99%
CAS No.: 149-57-5
Molecular Formula: C8H16O2
Formula Weight: 144.21
Flash Point: 114°(237°F)
Refractive Index: 1.4250
Melting Point: -59°
Boiling Point: 226-229°
Beilstein Reference: 1750468
MDL No.: MFCD00002675
Formula:
C₈H₁₆O₂ Boiling Pt: 226…229 °C Melting Pt: –59 °C Density: 0.91 Flash Pt: 114 °C (237 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002675 CAS Number: 149-57-5 EINECS: 205-743-6 |