1-Iodo-2,2-dimethylpropane, Sigma-Aldrich®
Supplier: Merck
Danger
301086-5G 301086-25G
SIAL301086-5GEA 90.5 EUR
SIAL301086-5G SIAL301086-25G
1-Iodo-2,2-dimethylpropane, Sigma-Aldrich®
1-Iodo-2,2-dimethylpropane
Formula: C₅H₁₁I
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.524 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 15501-33-4
EINECS: 239-533-0
UN: 1993
ADR: 3,III