3-Methoxy-3-methyl-1-butanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Methyl-3-methoxybutanol, Solvant 39, MMB, 3-Methoxy-3-methylbutan-1-ol
M-Clarity™ quality level = MQ100
408069-1L 408069-250ML
SIAL408069-1LEA 142 EUR
SIAL408069-1L SIAL408069-250ML
3-Methoxy-3-methyl-1-butanol, Sigma-Aldrich®
3-Methoxy-3-methyl-1-butanol
Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 174
Melting Pt: –50 °C
Density: 0.9038 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00044771
CAS Number: 56539-66-3
EINECS: 260-252-4