2,3-Dimethyl-2-butanol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
129240-25G
129240-5G
SIAL129240-25GEA
560
EUR
SIAL129240-25G
SIAL129240-5G
2,3-Dimethyl-2-butanol, Sigma-Aldrich®
2,3-Dimethyl-2-butanol
Formula:
C₆H₁₄O MW: 102.18 g/mol Boiling Pt: 119 Melting Pt: –10.4 °C Density: 0.8387 g/cm³ (0 °C) |
MDL Number:
MFCD00004463 CAS Number: 594-60-5 EINECS: 209-847-2 UN: 1987 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...