Isobutanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
M-Clarity™ quality level = MQ300
W217905-20KG-K W217905-2KG-K W217905-8KG-K
SIALW217905-20KG-KEA 371 EUR
SIALW217905-20KG-K SIALW217905-2KG-K SIALW217905-8KG-K
Isobutanol, Sigma-Aldrich®
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107.9
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3 III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM