tert-Butanol

Supplier: Roth Carl
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

4323.1 4323.2 4323.3
ROTH4323.1EA 0 EUR
ROTH4323.1 ROTH4323.2 ROTH4323.3
tert-Butanol
tert-Butanol
Formula: (CH₃)₃COH
Boiling Pt: 82.2 °C (1013 hPa)
Melting Pt: 25.3 °C
Density: 0.7887 g/cm³ (20 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3,II
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR