2-Methylhexane, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

M49704-5G M49704-25G
SIALM49704-5GEA 143 EUR
SIALM49704-5G SIALM49704-25G
2-Methylhexane, Sigma-Aldrich®
2-Methylhexane
Formula: C₇H₁₆
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: –118 °C
Density: 0.6793 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009487
CAS Number: 591-76-4
EINECS: 209-730-6
UN: 1206
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR