3-Methylhexane, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

M49801-25G M49801-5G
SIALM49801-25GEA 376 EUR
SIALM49801-25G SIALM49801-5G
3-Methylhexane, Sigma-Aldrich®
3-Methylhexane
Formula: C₇H₁₆
Boiling Pt: 92 °C (1013 hPa)
Melting Pt: –119.4 °C
Density: 0.687 g/cm³ (21 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009408
CAS Number: 589-34-4
EINECS: 209-643-3
UN: 1206
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR