4-(Indol-3-yl)butyric acid 98%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 3-Indolebutyric acid, Indole-3-butyric acid
160690050 160690250 160691000
ACRO160690050EA 78.9 EUR
ACRO160690050 ACRO160690250 ACRO160691000
4-(Indol-3-yl)butyric acid 98%
4-(Indol-3-yl)butyric acid
Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Melting Pt: 120…125 °C
Density: 1.27
Storage Temperature: Refrigerator
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,04987
Order Now
Specification Test Results
Appearance White to yellow to beige to brown Powder
Infrared spectrum Conforms
Melting point 120 to 125 °C
Titration with NaOH ≥97.5 %