4-(Indol-3-yl)butyric acid
Supplier: Molekula
Synonyms: 3-Indolebutyric acid, Indole-3-butyric acid
26194554-25G 26194554-5G
MOLE26194554-25GEA 98 EUR
MOLE26194554-25G MOLE26194554-5G
4-(Indol-3-yl)butyric acid
4-(Indol-3-yl)butyric acid
Formula: C₁₂H₁₃NO₂
MW: 203.24 g/mol
Melting Pt: 121…126 °C
Density: 1.27 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005664
CAS Number: 133-32-4
EINECS: 205-101-5
UN: 2811
ADR: 6.1 III
Merck Index: 13,04987