Ethyl phenylacetate ≥99%
Supplier: Thermo Fisher Scientific
Warning
A19356.0E A19356.22 A19356.36
A19356.0EEA 200 EUR
A19356.0E A19356.22 A19356.36
Ethyl phenylacetate ≥99%
Ethyl phenylacetate
Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 223…225 °C
Melting Pt: –29 °C
Density: 1.030
Flash Pt: 77 °C (170 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009178
CAS Number: 101-97-3
EINECS: 202-993-8
Merck Index: 14,3840