3-Phenylpropionic acid, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-Phenylpropanoic acid, Hydrocinnamic acid
M-Clarity™ quality level = MQ300
W288918-100G-K W288918-10KG-K W288918-1KG-K
SIALW288918-100G-KEA 77.8 EUR
SIALW288918-100G-K SIALW288918-10KG-K SIALW288918-1KG-K
3-Phenylpropionic acid, Sigma-Aldrich®
3-Phenylpropionic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Boiling Pt: 280
Melting Pt: 46…47 °C
Density: 1.071 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002771
CAS Number: 501-52-0
EINECS: 207-924-5
Merck Index: 13,04805