Phenylacetic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
P16621-100G P16621-500G P16621-5G
SIALP16621-100GEA 42.5 EUR
SIALP16621-100G SIALP16621-500G SIALP16621-5G
Phenylacetic acid, Sigma-Aldrich®
Phenylacetic acid
Formula: C₈H₈O₂
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 74…77 °C
Density: 1.081 g/cm³ (20 °C)
Flash Pt: 132 °C
MDL Number: MFCD00004313
CAS Number: 103-82-2
EINECS: 203-148-6
Merck Index: 12,07422